This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh04958957_g1
See other EEF1G GE Assays ›
Gene Symbol:
EEF1G
Gene Name:
eukaryotic translation elongation factor 1 gamma
Gene Aliases:
-
Chromosome Location:
Chr.14: 11629434 - 11641998 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh04958957_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: EEF1G
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001195580.1 NP_001182509.1 180 68
XM_028832882.1 XP_028688715.1 176 68
GenBank mRNA DR772456.1 - 184 68
JU327499.1 - 153 68
JU481838.1 - 279 68
JV043201.1 - 215 68

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_028832882.1 transcripts.

Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Macaca mulatta

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-cjlz8:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0