This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh05018261_m1
See other MIER1 GE Assays ›
Gene Symbol:
MIER1
Gene Name:
MIER1 transcriptional regulator
Gene Aliases:
EGK_00859
Chromosome Location:
Chr.1: 157446149 - 157510683 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh05018261_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MIER1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_015141830.2 XP_014997316.1 559 84
XM_015141845.2 XP_014997331.1 450 84
XM_015141849.2 XP_014997335.1 665 84
XM_015141853.2 XP_014997339.1 633 84
XM_015141859.2 XP_014997345.1 685 84
XM_015141867.2 XP_014997353.1 688 84
XM_015141876.2 XP_014997362.1 692 84
XM_015141884.2 XP_014997370.1 576 84
XM_015141889.2 XP_014997375.1 707 84
XM_015141894.2 XP_014997380.1 650 84
XM_015141904.2 XP_014997390.1 484 84
XM_015141915.2 XP_014997401.1 450 84
XM_028832757.1 XP_028688590.1 651 84
GenBank mRNA JU320719.1 - 444 84
JU320720.1 - 534 84
JU476575.1 - 658 84
JU476576.1 - 471 84
JV045415.1 - 455 84

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Macaca mulatta

Panther Classification:

Molecular Function -

homeodomain transcription factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0