This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh05927981_m1
See other CECR2 GE Assays ›
Gene Symbol:
CECR2
Gene Name:
CECR2 histone acetyl-lysine reader
Gene Aliases:
-
Chromosome Location:
Chr.10: 34459692 - 34657792 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh05927981_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CECR2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_028828020.1 XP_028683853.1 637 62
XM_028828021.1 XP_028683854.1 240 62
XM_028828022.1 XP_028683855.1 262 62
XM_028828023.1 XP_028683856.1 237 62
XM_028828024.1 XP_028683857.1 238 62
XM_028828025.1 XP_028683858.1 295 62
GenBank mRNA JV648591.1 - 212 62

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_028828025.1 transcripts.

Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Macaca mulatta

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-hwq2n:80/100.66.77.213:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0