This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh05944461_m1
See other ALDH3B1 GE Assays ›
Gene Symbol:
ALDH3B1
Gene Name:
aldehyde dehydrogenase 3 family member B1
Gene Aliases:
-
Chromosome Location:
Chr.14: 6750769 - 6775932 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh05944461_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ALDH3B1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_001103584.4 XP_001103584.2 371 73
XM_001103842.4 XP_001103842.1 457 73
XM_015113510.2 XP_014968996.1 518 73
XM_015113514.2 XP_014969000.1 431 73
XM_028833147.1 XP_028688980.1 612 73
XM_028833148.1 XP_028688981.1 785 73
XM_028833149.1 XP_028688982.1 368 73
XM_028833150.1 XP_028688983.1 370 73
GenBank mRNA JU325908.1 - 225 73

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_028833149.1, XM_028833150.1 transcripts.

Set Membership:

Amplicon length between 71 and 85 Recommended Assay Macaca mulatta

Panther Classification:

Molecular Function -

dehydrogenase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-7dtsd:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0