This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh05946982_g1
See other METTL26 GE Assays ›
Gene Symbol:
METTL26
Gene Name:
methyltransferase like 26
Gene Aliases:
-
Chromosome Location:
Chr.20: 662310 - 664168 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh05946982_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: METTL26
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001204163.2 NP_001191092.1 197 77
XM_015125162.2 XP_014980648.1 288 77
XM_015125163.2 XP_014980649.1 287 77
XM_028840389.1 XP_028696222.1 287 77
XM_028840390.1 XP_028696223.1 286 77
GenBank mRNA JU336713.1 - 252 77
JV048445.1 - 232 77
JV669632.1 - 225 77

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Recommended Assay Macaca mulatta

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0