Assay Details
| Gene Symbol: CTSS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015151744.2 | XP_015007230.1 | 1042 | 75 | ||
| XM_028829281.1 | XP_028685114.1 | 996 | 75 | |||
| GenBank mRNA | JU333113.1 | - | 1025 | 75 | ||
| JU474445.1 | - | 1018 | 75 | |||
| JU474446.1 | - | 1018 | 75 | |||
| JU474447.1 | - | 1018 | 75 | |||
| JU474448.1 | - | 1018 | 75 | |||
| JU474449.1 | - | 1018 | 75 | |||
More Information
Set Membership: |
|

CTSS
Genomic Map