This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh07215727_m1
See other TMEM218 GE Assays ›
Gene Symbol:
TMEM218
Gene Name:
transmembrane protein 218
Gene Aliases:
EGK_07055
Chromosome Location:
Chr.14: 117872376 - 117887170 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh07215727_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TMEM218
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_015116045.2 XP_014971531.1 612 81
XM_015116046.2 XP_014971532.1 602 81
XM_015116047.2 XP_014971533.1 647 81
XM_015116049.2 XP_014971535.1 569 81
XM_015116050.2 XP_014971536.1 627 81
XM_015116052.2 XP_014971538.1 716 81
XM_015116054.2 XP_014971540.1 617 81
XM_015116055.2 XP_014971541.1 600 81
XM_028834132.1 XP_028689965.1 431 81
XM_028834134.1 XP_028689967.1 577 81
XM_028834135.1 XP_028689968.1 1033 81
XM_028834136.1 XP_028689969.1 567 81
XM_028834137.1 XP_028689970.1 615 81
GenBank mRNA JU340315.1 - 197 81
JU473622.1 - 244 81
JV046446.1 - 236 81

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Recommended Assay Macaca mulatta

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-m2kmk:80/100.66.77.245:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0