Assay Details
| Gene Symbol: NEURL1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002804637.3 | XP_002804683.1 | 1597 | 105 | ||
| GenBank mRNA | JV600748.1 | - | 1569 | 105 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

NEURL1B| Gene Symbol: NEURL1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002804637.3 | XP_002804683.1 | 1597 | 105 | ||
| GenBank mRNA | JV600748.1 | - | 1569 | 105 | ||
Set Membership: |
|