This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh07217232_g1
See other GCNT1 GE Assays ›
Gene Symbol:
GCNT1
Gene Name:
glucosaminyl (N-acetyl) transferase 1
Gene Aliases:
EGK_07799
Chromosome Location:
Chr.15: 91807594 - 91916216 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh07217232_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GCNT1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_015117920.2 XP_014973406.1 419 99
XM_015117921.2 XP_014973407.1 603 99
XM_015117922.2 XP_014973408.1 270 99
XM_028835260.1 XP_028691093.1 187 99
XM_028835261.1 XP_028691094.1 228 99
XM_028835262.1 XP_028691095.1 418 99
GenBank mRNA JV711890.1 - 104 99

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_015117920.2, XM_015117921.2, XM_015117922.2, XM_028835260.1, XM_028835261.1, XM_028835262.1 transcripts.

Set Membership:

Amplicon length between 86 and 100 3' Most Macaca mulatta

Panther Classification:

Molecular Function -

glycosyltransferase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0