This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh07218619_g1
See other RPUSD1 GE Assays ›
Gene Symbol:
RPUSD1
Gene Name:
RNA pseudouridine synthase domain containing 1
Gene Aliases:
EGK_12314
Chromosome Location:
Chr.20: 814815 - 818185 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh07218619_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RPUSD1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_015125245.2 XP_014980731.2 568 84
XM_028840607.1 XP_028696440.1 485 84
GenBank mRNA JU332329.1 - 537 84
JU474188.1 - 533 84
JV567813.1 - 541 84
JV604799.1 - 537 84

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_028840607.1 transcripts.

Set Membership:

Amplicon length between 71 and 85 Recommended Assay Macaca mulatta

Panther Classification:

Molecular Function -

RNA processing factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0