This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh07218819_m1
See other NAT1 GE Assays ›
Gene Symbol:
NAT1
Gene Name:
N-acetyltransferase 1
Gene Aliases:
EGK_18730
Chromosome Location:
Chr.8: 18359603 - 18373413 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh07218819_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NAT1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_001098437.4 XP_001098437.1 475 102
XM_015144866.2 XP_015000352.1 856 102
XM_028852418.1 XP_028708251.1 693 102
XM_028852419.1 XP_028708252.1 731 102
XM_028852420.1 XP_028708253.1 672 102
GenBank mRNA JU495814.1 - 370 102

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_001098437.4, XM_015144866.2, XM_028852418.1, XM_028852419.1, XM_028852420.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 3' Most Macaca mulatta

Panther Classification:

Molecular Function -

acetyltransferase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0