This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh07219021_g1
See other SLC35B2 GE Assays ›
Gene Symbol:
SLC35B2
Gene Name:
solute carrier family 35 member B2
Gene Aliases:
-
Chromosome Location:
Chr.4: 125072386 - 125075868 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh07219021_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SLC35B2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_015136445.2 XP_014991931.1 521 87
GenBank mRNA JU320233.1 - 465 87
JU471876.1 - 487 87
JV044776.1 - 477 87
JV600190.1 - 467 87

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Recommended Assay Macaca mulatta

Panther Classification:

Molecular Function -

secondary carrier transporter

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-lsr54:80/100.66.79.61:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0