This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh07219995_g1
See other POLR3G GE Assays ›
Gene Symbol:
POLR3G
Gene Name:
RNA polymerase III subunit G
Gene Aliases:
-
Chromosome Location:
Chr.6: 87343716 - 87385189 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh07219995_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: POLR3G
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_002804454.3 XP_002804500.2 983 91
XM_002804455.3 XP_002804501.2 717 91
XM_015140393.2 XP_014995879.2 727 91
XM_015140394.2 XP_014995880.2 993 91
XM_015140395.2 XP_014995881.2 735 91
XM_015140396.2 XP_014995882.2 980 91
XM_028849571.1 XP_028705404.1 735 91
XM_028849572.1 XP_028705405.1 990 91
XM_028849573.1 XP_028705406.1 942 91
GenBank mRNA JU332556.1 - 778 91
JU474027.1 - 640 91
JV045956.1 - 696 91

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Recommended Assay Macaca mulatta

Panther Classification:

Molecular Function -

DNA-directed RNA polymerase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-d7bxm:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0