This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh07221141_g1
See other YTHDF3 GE Assays ›
Gene Symbol:
YTHDF3
Gene Name:
YTH N6-methyladenosine RNA binding protein 3
Gene Aliases:
-
Chromosome Location:
Chr.8: 63743527 - 63785980 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh07221141_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: YTHDF3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_002808481.3 XP_002808527.2 300 119
XM_015145449.2 XP_015000935.1 613 119
XM_015145450.2 XP_015000936.1 455 119
XM_015145452.2 XP_015000938.1 416 119
XM_028852697.1 XP_028708530.1 255 119
XM_028852698.1 XP_028708531.1 149 119
GenBank mRNA JU333794.1 - 421 119
JU475029.1 - 460 119
JV046649.1 - 462 119
JV647322.1 - 466 119

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_002808481.3, XM_028852698.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Recommended Assay Macaca mulatta

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0