This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh07222079_m1
See other MOB2 GE Assays ›
Gene Symbol:
MOB2
Gene Name:
MOB kinase activator 2
Gene Aliases:
EGK_05850
Chromosome Location:
Chr.14: 1525527 - 1597714 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh07222079_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MOB2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_015113321.2 XP_014968807.1 380 90
XM_015113322.2 XP_014968808.1 369 90
XM_015113323.2 XP_014968809.1 380 90
XM_028833009.1 XP_028688842.1 323 90
XM_028833010.1 XP_028688843.1 620 90
GenBank mRNA JU327042.1 - 83 90
JU477351.1 - 275 90
JV635860.1 - 326 90

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_028833010.1 transcripts.

Set Membership:

Amplicon length between 86 and 100 3' Most Recommended Assay Macaca mulatta

Panther Classification:

Molecular Function -

kinase activator

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-gv9bp:80/100.66.76.113:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline