This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh07225718_m1
See other UBE3D GE Assays ›
Gene Symbol:
UBE3D
Gene Name:
ubiquitin protein ligase E3D
Gene Aliases:
EGK_15114, UBE2CBP
Chromosome Location:
Chr.4: 90040534 - 90226508 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh07225718_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: UBE3D
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_002808392.3 XP_002808438.3 965 73
XM_015136839.2 XP_014992325.1 947 73
XM_028847392.1 XP_028703225.1 965 73
XM_028847393.1 XP_028703226.1 947 73
XM_028847394.1 XP_028703227.1 947 73
XM_028847395.1 XP_028703228.1 871 73
XM_028847396.1 XP_028703229.1 880 73
GenBank mRNA JU336984.1 - 892 73

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Recommended Assay Macaca mulatta

Panther Classification:

Molecular Function -

ubiquitin-protein ligase protein modifying enzyme

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0