This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh07227991_s1
See other RTP1 GE Assays ›
Gene Symbol:
RTP1
Gene Name:
receptor transporter protein 1
Gene Aliases:
-
Chromosome Location:
Chr.2: 10626646 - 10688670 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh07227991_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RTP1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_001103282.3 XP_001103282.1 1928 93
XM_015132436.2 XP_014987922.1 1495 93
XM_015132437.2 XP_014987923.1 1368 93
GenBank mRNA JV494983.1 - 521 93

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Macaca mulatta

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2tqsm:80/100.66.78.74:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0