This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh07228319_g1
See other MPI GE Assays ›
Gene Symbol:
MPI
Gene Name:
mannose phosphate isomerase
Gene Aliases:
EGK_17681
Chromosome Location:
Chr.7: 51932999 - 51941893 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh07228319_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MPI
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_015142733.2 XP_014998219.1 1289 64
XM_015142734.2 XP_014998220.1 822 64
XM_015142735.2 XP_014998221.1 885 64
XM_015142737.2 XP_014998223.2 774 64
XM_028851141.1 XP_028706974.1 794 64
XM_028851142.1 XP_028706975.1 901 64
XM_028851143.1 XP_028706976.1 772 64
GenBank mRNA JU334576.1 - 684 64
JU475660.1 - 688 64
JV651909.1 - 703 64

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Macaca mulatta

Panther Classification:

Molecular Function -

isomerase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0