This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh07232768_s1
See other TAL1 GE Assays ›
Gene Symbol:
TAL1
Gene Name:
TAL bHLH transcription factor 1, erythroid differentiation factor
Gene Aliases:
-
Chromosome Location:
Chr.1: 177271872 - 177288460 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh07232768_s1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TAL1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_015138968.2 XP_014994454.1 3909 77
XM_015138971.2 XP_014994457.1 3194 77
XM_028834022.1 XP_028689855.1 4502 77
XM_028834026.1 XP_028689859.1 3612 77
XM_028834032.1 XP_028689865.1 2446 77
GenBank mRNA JV722050.1 - 1623 77

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Recommended Assay Macaca mulatta

Panther Classification:

Molecular Function -

basic helix-loop-helix transcription factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-gxkmn:80/100.66.78.214:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0