This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh07232899_m1
See other EEF1AKMT1 GE Assays ›
Gene Symbol:
EEF1AKMT1
Gene Name:
EEF1A lysine methyltransferase 1
Gene Aliases:
EGK_09149
Chromosome Location:
Chr.17: 400321 - 451971 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh07232899_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: EEF1AKMT1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_015120704.2 XP_014976190.1 888 96
XM_028837490.1 XP_028693323.1 888 96
XM_028837491.1 XP_028693324.1 240 96
XM_028837492.1 XP_028693325.1 250 96
XM_028837493.1 XP_028693326.1 888 96
XM_028837494.1 XP_028693327.1 240 96
XM_028837495.1 XP_028693328.1 264 96
XM_028837496.1 XP_028693329.1 888 96
XM_028837497.1 XP_028693330.1 238 96
XM_028837498.1 XP_028693331.1 250 96
GenBank mRNA JU323153.1 - 213 96
JU473625.1 - 190 96
JV054780.1 - 208 96
JV667218.1 - 183 96

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Recommended Assay Macaca mulatta

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-wt24k:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline