This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh07235484_s1
See other LOC720244 GE Assays ›
Gene Symbol:
LOC720244
Gene Name:
zinc finger protein 83
Gene Aliases:
ZNF83
Chromosome Location:
Chr.19: 52282357 - 52576657 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh07235484_s1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC720244
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_015124769.2 XP_014980255.1 2550 82
XM_015124771.2 XP_014980257.1 3400 82
XM_015124772.2 XP_014980258.1 2379 82
XM_028839396.1 XP_028695229.1 2575 82
XM_028839397.1 XP_028695230.1 3074 82
XM_028839398.1 XP_028695231.1 2498 82
XM_028839399.1 XP_028695232.1 2490 82
XM_028839400.1 XP_028695233.1 2490 82
GenBank mRNA JV718632.1 - 718 82

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Recommended Assay Macaca mulatta

Panther Classification:

Molecular Function -

C2H2 zinc finger transcription factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-nwsk8:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0