This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh07236133_s1
See other IFIT3 GE Assays ›
Gene Symbol:
IFIT3
Gene Name:
interferon induced protein with tetratricopeptide repeats 3
Gene Aliases:
-
Chromosome Location:
Chr.9: 89585075 - 89596574 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh07236133_s1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: IFIT3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_001086192.4 XP_001086192.2 853 85
XM_028826632.1 XP_028682465.1 811 85
GenBank mRNA JU332327.1 - 706 85
JU471163.1 - 706 85
JU471164.1 - 686 85
JV045391.1 - 700 85
JV580534.1 - 724 85

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Recommended Assay Macaca mulatta

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0