This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn00493446_m1
See other TCF7 GE Assays ›
Gene Symbol:
Tcf7
Gene Name:
transcription factor 7 (T-cell specific, HMG-box)
Gene Aliases:
-
Chromosome Location:
Chr.10: 37616022 - 37647851 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn00493446_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price 2,694.00
Your Price
Online offer:
2694.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tcf7
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_006246362.2 XP_006246424.1 4 - 5 784 88
XM_006246363.2 XP_006246425.1 4 - 5 794 88
XM_006246364.2 XP_006246426.1 4 - 5 788 88
XM_006246365.2 XP_006246427.1 4 - 5 784 88
XM_008767741.1 XP_008765963.1 4 - 5 2608 88

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Recommended Assay Rattus norvegicus Inventoried Probe spans exons

Panther Classification:

Molecular Function -

DNA-binding transcription factor

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
brain development
biological_process
Wnt signaling pathway
neural tube development
embryonic genitalia morphogenesis
T cell receptor V(D)J recombination
regulation of cell proliferation
canonical Wnt signaling pathway involved in negative regulation of apoptotic process
alpha-beta T cell differentiation
embryonic digestive tract morphogenesis
embryonic hindgut morphogenesis
generation of neurons
canonical Wnt signaling pathway
cellular response to interleukin-4
core promoter binding
transcriptional repressor activity, RNA polymerase II transcription regulatory region sequence-specific binding
molecular_function
DNA binding
chromatin binding
transcription factor activity, sequence-specific DNA binding
beta-catenin binding
sequence-specific DNA binding
transcription regulatory region DNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0