This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn00509060_m1
See other L3HYPDH GE Assays ›
Gene Symbol:
L3hypdh
Gene Name:
L-3-hydroxyproline dehydratase (trans-)
Gene Aliases:
RGD1305721
Chromosome Location:
Chr.6: 94824112 - 94834908 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn00509060_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: L3hypdh
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001108031.1 NP_001101501.1 1 - 2 813 56

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0600002635.rn.1

Set Membership:

Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons

Panther Classification:

Molecular Function -

dehydratase

Gene Ontology Categories:

Function(s) Process(es)

metabolic process
hydro-lyase activity
proline racemase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0600018733.rn.1 TC0600002635.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0600018734.rn.1 TC0600002635.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-d7bxm:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0