This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn00563117_m1
See other AANAT GE Assays ›
Gene Symbol:
Aanat
Gene Name:
aralkylamine N-acetyltransferase
Gene Aliases:
AA-NAT, Nat4
Chromosome Location:
Chr.10: 105568091 - 105572407 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn00563117_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Aanat
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_012818.2 NP_036950.1 1 - 2 53 64
GenBank mRNA DQ075321.1 - 1 - 2 241 64
U38306.1 - 1 - 2 53 64
U40803.1 - 1 - 2 140 64

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the DQ075321.1, NM_012818.2, U38306.1, U40803.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Rattus norvegicus Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Aanat

Panther Classification:

Molecular Function -

acetyltransferase

Gene Ontology Categories:

Function(s) Process(es)

N-terminal protein amino acid acetylation
circadian rhythm
response to light stimulus
photoperiodism
response to zinc ion
response to organic cyclic compound
melatonin biosynthetic process
response to insulin
response to cytokine
response to prostaglandin E
entrainment of circadian clock by photoperiod
response to copper ion
response to corticosterone
response to cAMP
response to calcium ion
cellular response to cAMP
aralkylamine N-acetyltransferase activity
arylamine N-acetyltransferase activity
14-3-3 protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR1000017827.rn.1 TC1000001978.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1000017824.rn.1 TC1000001978.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-hwq2n:80/100.66.77.213:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0