This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn00563499_m1
See other TNFRSF11B GE Assays ›
Gene Symbol:
Tnfrsf11b
Gene Name:
tumor necrosis factor receptor superfamily, member 11b
Gene Aliases:
Opg
Chromosome Location:
Chr.7: 93798580 - 93826586 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn00563499_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information
  • Citations (1)

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tnfrsf11b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_012870.2 NP_037002.1 1 - 2 133 75
GenBank mRNA BC081830.1 - 1 - 2 133 7096099 75
U94330.1 - 1 - 2 152 75

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the BC081830.1, NM_012870.2, U94330.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Tnfrsf11b

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

apoptotic process
signal transduction
response to nutrient
response to inorganic substance
extracellular matrix organization
response to magnesium ion
negative regulation of odontogenesis of dentin-containing tooth
response to drug
response to estrogen
negative regulation of bone resorption
response to arsenic-containing substance
molecular_function
cytokine activity

MicroArray Probes:

ABI

Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 20929372 - -

Back To Top
 

Citations (1)

Catch-up growth after hypothyroidism is caused by delayed growth plate senescence.
Authors
Marino R, Hegde A, Barnes KM, Schrier L, Emons JA, Nilsson O, Baron J
Journal
Endocrinology - Page No 1820 (Tue Apr 01 00:00:00 EDT 2008)
Volume (Year)
149 (2008)
Product Usage
-

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-t8jrk:80/100.66.77.131:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0