This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn00571296_m1
See other BET1L GE Assays ›
Gene Symbol:
Bet1l
Gene Name:
Bet1 golgi vesicular membrane trafficking protein-like
Gene Aliases:
Bet1, Gs15
Chromosome Location:
Chr.1: 213602812 - 213606500 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn00571296_m1
Size
Availability Inventoried
Catalog # 4331182
Price 2,694.00
Your Price
Online offer:
2694.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Bet1l
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_019368.3 NP_062241.2 1 - 2 70 77
XM_006230574.2 XP_006230636.1 2 - 3 282 77
GenBank mRNA AF003998.1 - 1 - 2 109 77
BC059138.1 - 1 - 2 74 6890305 77
FQ228853.1 - 2 - 3 232 77

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AF003998.1, BC059138.1, NM_019368.3, XM_006230574.2 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Bet1l

Panther Classification:

Molecular Function -

SNARE protein

Gene Ontology Categories:

Function(s) Process(es)

protein transport
retrograde transport, endosome to Golgi
membrane fusion
regulation of retrograde vesicle-mediated transport, Golgi to ER
SNAP receptor activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0100054392.rn.1 TC0100006910.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0100054394.rn.1 TC0100006910.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0