This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn00571298_g1
See other BET1L GE Assays ›
Gene Symbol:
Bet1l
Gene Name:
Bet1 golgi vesicular membrane trafficking protein-like
Gene Aliases:
Bet1, Gs15
Chromosome Location:
Chr.1: 213602812 - 213606500 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn00571298_g1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Bet1l
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_019368.3 NP_062241.2 3 - 4 215 66
XM_006230574.2 XP_006230636.1 4 - 5 427 66
GenBank mRNA AF003998.1 - 3 - 4 254 66
BC059138.1 - 3 - 4 219 6890305 66
FQ228853.1 - 4 - 5 377 66

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0100006910.rn.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Bet1l

Panther Classification:

Molecular Function -

SNARE protein

Gene Ontology Categories:

Function(s) Process(es)

protein transport
retrograde transport, endosome to Golgi
membrane fusion
regulation of retrograde vesicle-mediated transport, Golgi to ER
SNAP receptor activity

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P346599 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 953984 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 922465 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 497438 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 146381 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P346599 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P346599 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P346599 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P158635 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P346599 - -
Matsumoto (Agilent Rat cDNA Microarray G4105A) 700230185 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 86846 - -
Agilent Rat cDNA Microarray (G4105A) 700230185 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10726656 - -
Affymetrix Rat Gene 1.0 ST Array 10726658 - -
Affymetrix Rat Gene 1.0 ST Array 10726657 - -
Affymetrix Rat Gene 1.0 ST Array 10726660 - -
Affymetrix Rat Gene 1.1 ST Array 10726655 - -
Rat Genome 230 2.0 Array 1372950_PM_at - -
Rat Genome 230 2.0 Array 1387448_PM_at - -
Rat Genome 230 2.0 Array 1372950_at - -
Rat Genome 230 2.0 Array 1387448_at - -
Affymetrix HT Rat Focus Array 1372950_PM_at - -
Affymetrix HT Rat Focus Array 1372950_at - -
Affymetrix HT Rat Focus Array 1387448_PM_at - -
Affymetrix HT Rat Focus Array 1387448_at - -
Affymetrix Rat Clariom S Assay PSR0100054389.rn.1 TC0100006910.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0100054390.rn.1 TC0100006910.rn.1 RTA TC
Rat Expression Set 230 1387448_PM_at - -
Rat Expression Set 230 1387448_at - -
Affymetrix Rat Gene 1.0 ST Array 10726655 - -
Rat Genome U34 Set AF003998_at - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 22070866 - -
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1650490 - -
RatRef-12 Expression BeadChip ILMN_55338 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0