This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn00574025_s1
See other GCNT1 GE Assays ›
Gene Symbol:
Gcnt1
Gene Name:
glucosaminyl (N-acetyl) transferase 1, core 2
Gene Aliases:
core 2 GlcNAc-T
Chromosome Location:
Chr.1: 236547734 - 236585037 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn00574025_s1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price 284.00
Your Price
Online offer: 266.65
266.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gcnt1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_022276.1 NP_071612.1 1 1852 88
XM_006231147.2 XP_006231209.1 3 1461 88
XM_006231150.2 XP_006231212.1 3 1553 88
GenBank mRNA S79797.1 - 1 1852 88

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gcnt1

Panther Classification:

Molecular Function -

glycosyltransferase

Gene Ontology Categories:

Function(s) Process(es)

glycoprotein biosynthetic process
response to insulin
tissue morphogenesis
leukocyte tethering or rolling
cell adhesion molecule production
kidney morphogenesis
beta-1,3-galactosyl-O-glycosyl-glycoprotein beta-1,6-N-acetylglucosaminyltransferase activity

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P497553 - -
Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P038462 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 923569 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 848918 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 756100 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 215355 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 500578 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P497553 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P038462 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P497553 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P038462 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P038462 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P497553 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P497553 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 31630 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10729210 - -
Affymetrix Rat Gene 1.0 ST Array 10729211 - -
Affymetrix Rat Gene 1.0 ST Array 10729212 - -
Affymetrix Rat Gene 1.1 ST Array 10729209 - -
Rat Genome 230 2.0 Array 1368770_PM_at - -
Rat Genome 230 2.0 Array 1368770_at - -
Affymetrix HT Rat Focus Array 1368770_at - -
Affymetrix HT Rat Focus Array 1368770_PM_at - -
Affymetrix Rat Clariom S Assay PSR0100027220.rn.1 TC0100003332.rn.1 RTA TC
Rat Expression Set 230 1368770_at - -
Rat Expression Set 230 1368770_PM_at - -
Affymetrix Rat Gene 1.0 ST Array 10729209 - -
Rat Genome U34 Set S79797_at - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 22327124 - -
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1649912 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0