This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn00578832_m1
See other ATF2 GE Assays ›
Gene Symbol:
Atf2
Gene Name:
activating transcription factor 2
Gene Aliases:
-
Chromosome Location:
Chr.3: 60721137 - 60795951 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn00578832_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Atf2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_031018.1 NP_112280.1 12 - 13 1470 96
GenBank mRNA AY724488.1 - 1048 96
M65148.1 - 7 - 8 865 96
U38938.1 - 12 - 13 1470 96

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0300003333.rn.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Recommended Assay Rattus norvegicus Rat Tumor Metastasis Panel 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Atf2

Panther Classification:

Molecular Function -

basic leucine zipper transcription factor

Gene Ontology Categories:

Function(s) Process(es)

outflow tract morphogenesis
transcription, DNA-templated
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
response to osmotic stress
cellular response to DNA damage stimulus
histone acetylation
intra-S DNA damage checkpoint
positive regulation of transforming growth factor beta2 production
fat cell differentiation
positive regulation of transcription from RNA polymerase II promoter
positive regulation of sequence-specific DNA binding transcription factor activity
adipose tissue development
positive regulation of mitochondrial membrane permeability involved in apoptotic process
RNA polymerase II regulatory region sequence-specific DNA binding
RNA polymerase II distal enhancer sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
RNA polymerase II activating transcription factor binding
enhancer sequence-specific DNA binding
DNA binding
chromatin binding
transcription factor activity, sequence-specific DNA binding
histone acetyltransferase activity
cAMP response element binding protein binding
protein kinase binding
cAMP response element binding
sequence-specific DNA binding
metal ion binding
protein heterodimerization activity

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P120589 - -
Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P316430 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 347462 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 257092 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 965143 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 291999 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 872124 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 468669 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 670757 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 139394 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 636915 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 216428 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 641815 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 152837 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 255245 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 496351 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 678874 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 50580 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 465617 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 641988 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 876267 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 792721 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 96302 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 800666 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 90975 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 543173 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 122522 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 179000 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 256127 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 740224 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 508962 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 629642 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 308515 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 114874 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 849066 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 837777 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 410844 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 839811 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 945910 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 820944 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 606833 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 911288 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 383575 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 287574 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 908012 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 624869 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 733244 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 705397 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 672643 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 422998 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 636804 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 203336 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 676253 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 120574 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 138706 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 1122 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 8796 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 122214 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 158891 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 143779 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P316430 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P120589 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P316430 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P120589 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P316430 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P120589 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P446409 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 92441 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 100351 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 69541 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 81468 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 65528 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10846237 - -
Affymetrix Rat Gene 1.0 ST Array 10846242 - -
Affymetrix Rat Gene 1.0 ST Array 10846246 - -
Affymetrix Rat Gene 1.0 ST Array 10846247 - -
Affymetrix Rat Gene 1.0 ST Array 10846238 - -
Affymetrix Rat Gene 1.0 ST Array 10846241 - -
Affymetrix Rat Gene 1.0 ST Array 10846244 - -
Affymetrix Rat Gene 1.0 ST Array 10846235 - -
Affymetrix Rat Gene 1.0 ST Array 10846239 - -
Affymetrix Rat Gene 1.0 ST Array 10846234 - -
Affymetrix Rat Gene 1.0 ST Array 10846240 - -
Affymetrix Rat Gene 1.0 ST Array 10846245 - -
Affymetrix Rat Gene 1.0 ST Array 10846236 - -
Affymetrix Rat Gene 1.0 ST Array 10846243 - -
Affymetrix Rat Gene 1.1 ST Array 10846233 - -
Rat Genome 230 2.0 Array 1369472_PM_a_at - -
Rat Genome 230 2.0 Array 1369472_a_at - -
Affymetrix HT Rat Focus Array 1369472_PM_a_at - -
Affymetrix HT Rat Focus Array 1369472_a_at - -
Affymetrix Rat Clariom S Assay PSR0300025872.rn.1 TC0300003333.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0300025873.rn.1 TC0300003333.rn.1 RTA TC
Rat Expression Set 230 1369472_PM_a_at - -
Rat Expression Set 230 1369472_a_at - -
Affymetrix Rat Gene 1.0 ST Array 10846233 - -
Rat Genome U34 Set U38938_i_at - -
Rat Genome U34 Set M65148_s_at - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 22319228 - -
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1376383 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2k58s:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0