This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn00583439_m1
See other PNLIPRP1 GE Assays ›
Gene Symbol:
Pnliprp1
Gene Name:
pancreatic lipase-related protein 1
Gene Aliases:
-
Chromosome Location:
Chr.1: 279867034 - 279882663 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn00583439_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price 2,694.00
Your Price
Online offer:
2694.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pnliprp1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_032081.1 NP_114470.1 10 - 11 1138 99
GenBank mRNA FQ232772.1 - 10 - 11 1138 99
X61925.1 - 10 - 11 1138 99

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0100003890.rn.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Pnliprp1

Panther Classification:

Molecular Function -

lipase

Gene Ontology Categories:

Function(s) Process(es)

lipid metabolic process
pancreas development
response to peptide hormone
response to glucocorticoid
triglyceride lipase activity
calcium ion binding
protein binding
lipase activity

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P386017 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 886484 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 902849 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 232694 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 465873 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 98386 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 300531 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 531348 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 876488 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 160588 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 682194 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 49581 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 310192 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 579129 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 117635 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 91412 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P386017 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P386017 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P386017 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P386017 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 7293 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10716397 - -
Affymetrix Rat Gene 1.0 ST Array 10716390 - -
Affymetrix Rat Gene 1.0 ST Array 10716392 - -
Affymetrix Rat Gene 1.0 ST Array 10716395 - -
Affymetrix Rat Gene 1.0 ST Array 10716391 - -
Affymetrix Rat Gene 1.0 ST Array 10716388 - -
Affymetrix Rat Gene 1.0 ST Array 10716396 - -
Affymetrix Rat Gene 1.0 ST Array 10716389 - -
Affymetrix Rat Gene 1.0 ST Array 10716394 - -
Affymetrix Rat Gene 1.0 ST Array 10716401 - -
Affymetrix Rat Gene 1.0 ST Array 10716400 - -
Affymetrix Rat Gene 1.0 ST Array 10716398 - -
Affymetrix Rat Gene 1.0 ST Array 10716399 - -
Affymetrix Rat Gene 1.0 ST Array 10716393 - -
Affymetrix Rat Gene 1.1 ST Array 10716387 - -
Rat Genome 230 2.0 Array 1368532_at - -
Rat Genome 230 2.0 Array 1368532_PM_at - -
Affymetrix HT Rat Focus Array 1368532_PM_at - -
Affymetrix HT Rat Focus Array 1368532_at - -
Affymetrix Rat Clariom S Assay PSR0100031822.rn.1 TC0100003890.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0100031826.rn.1 TC0100003890.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0100031820.rn.1 TC0100003890.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0100031824.rn.1 TC0100003890.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0100031818.rn.1 TC0100003890.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0100031817.rn.1 TC0100003890.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0100031825.rn.1 TC0100003890.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0100031815.rn.1 TC0100003890.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0100031827.rn.1 TC0100003890.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0100031819.rn.1 TC0100003890.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0100031821.rn.1 TC0100003890.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0100031816.rn.1 TC0100003890.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0100031823.rn.1 TC0100003890.rn.1 RTA TC
Rat Expression Set 230 1368532_PM_at - -
Rat Expression Set 230 1368532_at - -
Affymetrix Rat Gene 1.0 ST Array 10716387 - -
Rat Genome U34 Set rc_AA801219_at - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 21271415 - -
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1361342 - -
RatRef-12 Expression BeadChip ILMN_57633 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-7dtsd:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0