This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn00583723_m1
See other ABCD2 GE Assays ›
Gene Symbol:
Abcd2
Gene Name:
ATP-binding cassette, subfamily D (ALD), member 2
Gene Aliases:
ALDR
Chromosome Location:
Chr.7: 132294564 - 132343169 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn00583723_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Abcd2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_033352.1 NP_203503.1 9 - 10 2205 99
GenBank mRNA AF131294.1 - 9 - 10 2205 99

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0700003724.rn.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Transporter 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Abcd2

Panther Classification:

Molecular Function -

ATP-binding cassette (ABC) transporter

Gene Ontology Categories:

Function(s) Process(es)

very long-chain fatty acid metabolic process
fatty acid beta-oxidation
metabolic process
positive regulation of fatty acid beta-oxidation
very long-chain fatty acid catabolic process
transmembrane transport
ATP binding
ATPase activity, coupled to transmembrane movement of substances
protein homodimerization activity

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P133967 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 940788 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 210829 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 478034 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 896442 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 577692 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 689059 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 105958 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 79291 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 528092 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 291980 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 660668 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 826901 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 837367 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 447119 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 69108 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 636035 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 682088 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 541053 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 228446 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 695076 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 213496 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 531713 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 772776 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 710898 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 943816 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 740706 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 728417 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 771983 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 176381 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 63321 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 878873 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 162134 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 369236 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 525790 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 203557 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 78859 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 158410 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 89231 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 166364 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 7321 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 30338 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P133967 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P133967 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P133967 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P525170 - -
Matsumoto (Agilent Rat cDNA Microarray G4105A) 700499870 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 69546 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 65091 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 84383 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 50245 - -
Agilent Rat cDNA Microarray (G4105A) 700499870 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10906432 - -
Affymetrix Rat Gene 1.0 ST Array 10906429 - -
Affymetrix Rat Gene 1.0 ST Array 10906438 - -
Affymetrix Rat Gene 1.0 ST Array 10906434 - -
Affymetrix Rat Gene 1.0 ST Array 10906433 - -
Affymetrix Rat Gene 1.0 ST Array 10906436 - -
Affymetrix Rat Gene 1.0 ST Array 10906430 - -
Affymetrix Rat Gene 1.0 ST Array 10906431 - -
Affymetrix Rat Gene 1.0 ST Array 10906437 - -
Affymetrix Rat Gene 1.0 ST Array 10906435 - -
Affymetrix Rat Gene 1.1 ST Array 10906428 - -
Rat Genome 230 2.0 Array 1368561_PM_at - -
Rat Genome 230 2.0 Array 1368561_at - -
Affymetrix HT Rat Focus Array 1368561_PM_at - -
Affymetrix HT Rat Focus Array 1368561_at - -
Affymetrix Rat Clariom S Assay PSR0700029768.rn.1 TC0700003724.rn.1 RTA TC
Rat Expression Set 230 1368561_PM_at - -
Rat Expression Set 230 1368561_at - -
Affymetrix Rat Gene 1.0 ST Array 10906428 - -
Rat Genome U34 Set rc_AI070167_at - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 21883525 - -
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1367993 - -
RatRef-12 Expression BeadChip ILMN_48309 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-55658555d6-7gxhj:80/100.66.79.224:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline