This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn00585530_m1
See other COX17 GE Assays ›
Gene Symbol:
Cox17
Gene Name:
COX17 cytochrome c oxidase copper chaperone
Gene Aliases:
-
Chromosome Location:
Chr.11: 64962663 - 64968437 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn00585530_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price 2,694.00
Your Price
Online offer:
2694.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cox17
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_053540.2 NP_445992.1 1 - 2 203 66
GenBank mRNA AB032178.1 - 1 - 2 147 66
BI293817.1 - 2 - 3 66
BQ195675.1 - 2 - 3 66

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC1100002052.rn.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cox17

Panther Classification:

Molecular Function -

chaperone

Gene Ontology Categories:

Function(s) Process(es)

copper ion transport
brain development
heart development
positive regulation of catalytic activity
molecular_function
copper ion binding
enzyme activator activity
copper chaperone activity

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P12844 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 746190 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 81078 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12844 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12844 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P12844 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12844 - -
Matsumoto (Agilent Rat cDNA Microarray G4105A) 700302103 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 41291 - -
Agilent Rat cDNA Microarray (G4105A) 700302103 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10754186 - -
Affymetrix Rat Gene 1.0 ST Array 10754187 - -
Affymetrix Rat Gene 1.1 ST Array 10754185 - -
Rat Genome 230 2.0 Array 1369984_at - -
Rat Genome 230 2.0 Array 1369984_PM_at - -
Affymetrix HT Rat Focus Array 1369984_PM_at - -
Affymetrix HT Rat Focus Array 1369984_at - -
Affymetrix Rat Clariom S Assay PSR1100013937.rn.1 TC1100002052.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1100013939.rn.1 TC1100002052.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1100013940.rn.1 TC1100002052.rn.1 RTA TC
Rat Expression Set 230 1369984_at - -
Rat Expression Set 230 1369984_PM_at - -
Affymetrix Rat Gene 1.0 ST Array 10754185 - -
Rat Genome U34 Set rc_AI138072_at - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 20967521 - -
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1353069 - -
RatRef-12 Expression BeadChip ILMN_59410 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0