This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn00586046_m1
See other NUPR1 GE Assays ›
Gene Symbol:
Nupr1 NEWGENE_69363
Gene Name:
nuclear protein, transcriptional regulator, 1
nuclear protein, transcriptional regulator, 1
Gene Aliases:
p8
Nupr1
Chromosome Location:
Chr.1: 194767484 - 194769519 on Build Rnor_6.0
Chromosome Location:
Chr.1: 198043117 - 198045203 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
-
Assay ID Rn00586046_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information
  • Citations (1)

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Nupr1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_053611.2 NP_446063.1 2 - 3 341 112
GenBank mRNA AF014503.1 - 2 - 3 309 112
Gene Symbol: NEWGENE_69363
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_008759924.1 XP_008758146.1 2 - 3 392 112

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis Rattus norvegicus 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Nupr1
Click here to find siRNAs for Gene NEWGENE_69363

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P812008 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 432505 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 93839 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P812008 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P812008 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P812008 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P812008 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 88857 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10725780 - -
Affymetrix Rat Gene 1.0 ST Array 10725781 - -
Affymetrix Rat Gene 1.0 ST Array 10725779 - -
Affymetrix Rat Gene 1.1 ST Array 10725778 - -
Rat Genome 230 2.0 Array 1367847_PM_at - -
Rat Genome 230 2.0 Array 1367847_at - -
Affymetrix HT Rat Focus Array 1367847_PM_at - -
Affymetrix HT Rat Focus Array 1367847_at - -
Affymetrix Rat Clariom S Assay PSR0100051651.rn.1 TC0100006586.rn.1 RTA TC
Rat Expression Set 230 1367847_at - -
Rat Expression Set 230 1367847_PM_at - -
Affymetrix Rat Gene 1.0 ST Array 10725778 - -
Rat Genome U34 Set AF014503_at - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 22345660 - -
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1354048 - -
RatRef-12 Expression BeadChip ILMN_52937 - -

Back To Top
 

Citations (1)

Catch-up growth after hypothyroidism is caused by delayed growth plate senescence.
Authors
Marino R, Hegde A, Barnes KM, Schrier L, Emons JA, Nilsson O, Baron J
Journal
Endocrinology - Page No 1820 (Tue Apr 01 00:00:00 EDT 2008)
Volume (Year)
149 (2008)
Product Usage
-

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-7gdsv:80/100.66.79.88:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0