This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn00589833_m1
See other NAPA GE Assays ›
Gene Symbol:
Napa
Gene Name:
N-ethylmaleimide-sensitive factor attachment protein, alpha
Gene Aliases:
alpha-SNAP
Chromosome Location:
Chr.1: 77994227 - 78012836 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn00589833_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price 284.00
Your Price
Online offer: 266.65
266.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Napa
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_080585.1 NP_542152.1 3 - 4 341 94
GenBank mRNA BC063156.1 - 3 - 4 400 6921187 94
FQ216375.1 - 3 - 4 659 94
X89968.1 - 3 - 4 341 94

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0100000921.rn.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Napa

Panther Classification:

Molecular Function -

membrane traffic protein

Gene Ontology Categories:

Function(s) Process(es)

intracellular protein transport
brain development
regulation of synaptic vesicle priming
neuron differentiation
synaptic transmission, glutamatergic
SNARE complex disassembly
apical protein localization
membrane fusion
SNARE binding
soluble NSF attachment protein activity
protein binding
syntaxin binding

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P262113 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 237972 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 683977 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 518383 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 473130 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 911883 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 698714 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 649056 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 939224 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 150347 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 145588 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 183881 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 762553 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 132505 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 150190 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 937283 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 818875 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 162995 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 82797 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P262113 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P262113 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P262113 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P262113 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 62406 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10704422 - -
Affymetrix Rat Gene 1.0 ST Array 10704421 - -
Affymetrix Rat Gene 1.0 ST Array 10704416 - -
Affymetrix Rat Gene 1.0 ST Array 10704419 - -
Affymetrix Rat Gene 1.0 ST Array 10704415 - -
Affymetrix Rat Gene 1.0 ST Array 10704414 - -
Affymetrix Rat Gene 1.0 ST Array 10704417 - -
Affymetrix Rat Gene 1.0 ST Array 10704420 - -
Affymetrix Rat Gene 1.0 ST Array 10704418 - -
Affymetrix Rat Gene 1.0 ST Array 10704423 - -
Affymetrix Rat Gene 1.1 ST Array 10704413 - -
Rat Genome 230 2.0 Array 1398782_at - -
Rat Genome 230 2.0 Array 1398782_PM_at - -
Affymetrix HT Rat Focus Array 1398782_at - -
Affymetrix HT Rat Focus Array 1398782_PM_at - -
Affymetrix Rat Clariom S Assay PSR0100006257.rn.1 TC0100000921.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0100006259.rn.1 TC0100000921.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0100006254.rn.1 TC0100000921.rn.1 RTA TC
Rat Expression Set 230 1398782_PM_at - -
Rat Expression Set 230 1398782_at - -
Affymetrix Rat Gene 1.0 ST Array 10704413 - -
Rat Genome U34 Set X89968_at - -
Rat Genome U34 Set X89968_g_at - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 22379409 - -
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1360892 - -
RatRef-12 Expression BeadChip ILMN_66723 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0