Assay Details
| Gene Symbol: Hadha | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130826.2 | NP_570839.2 | 13 - 14 | 1410 | 71 | |
| GenBank mRNA | BC091697.1 | - | 13 - 14 | 1410 | 7301707 | 71 |
| D16478.1 | - | 13 - 14 | 1408 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
fatty acid beta-oxidation
response to insulin
response to drug
oxidation-reduction process
fatty-acyl-CoA binding
3-hydroxyacyl-CoA dehydrogenase activity
acetyl-CoA C-acyltransferase activity
enoyl-CoA hydratase activity
long-chain-enoyl-CoA hydratase activity
long-chain-3-hydroxyacyl-CoA dehydrogenase activity
protein complex binding
NAD binding
fatty acid beta-oxidation
response to insulin
response to drug
oxidation-reduction process
fatty-acyl-CoA binding
3-hydroxyacyl-CoA dehydrogenase activity
acetyl-CoA C-acyltransferase activity
enoyl-CoA hydratase activity
long-chain-enoyl-CoA hydratase activity
long-chain-3-hydroxyacyl-CoA dehydrogenase activity
protein complex binding
NAD binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P055984 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P480296 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 30710 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 910163 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 661066 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 389625 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 843629 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 173646 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 110971 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 286568 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 505745 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 66043 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 114245 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P055984 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P480296 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P055984 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P480296 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P055984 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P480296 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P400202 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P480296 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P414469 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 81400 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 103977 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 66225 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 85794 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10883308 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10883309 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10883304 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10883307 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10883306 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10883310 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10883305 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10883303 | - | - |
| Rat Genome 230 2.0 Array | 1394763_at | - | - |
| Rat Genome 230 2.0 Array | 1394857_at | - | - |
| Rat Genome 230 2.0 Array | 1394857_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1394763_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1370164_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1370164_at | - | - |
| Affymetrix HT Rat Focus Array | 1394857_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1394857_at | - | - |
| Affymetrix HT Rat Focus Array | 1394763_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1370164_at | - | - |
| Affymetrix HT Rat Focus Array | 1370164_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1394763_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0600002146.rn.1 | TC0600000317.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0600002149.rn.1 | TC0600000317.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0600002140.rn.1 | TC0600000317.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0600002141.rn.1 | TC0600000317.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0600002142.rn.1 | TC0600000317.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0600002151.rn.1 | TC0600000317.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0600002155.rn.1 | TC0600000317.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0600002156.rn.1 | TC0600000317.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0600002158.rn.1 | TC0600000317.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0600002144.rn.1 | TC0600000317.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0600002154.rn.1 | TC0600000317.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0600002145.rn.1 | TC0600000317.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0600002150.rn.1 | TC0600000317.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0600002152.rn.1 | TC0600000317.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0600002153.rn.1 | TC0600000317.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0600002138.rn.1 | TC0600000317.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0600002143.rn.1 | TC0600000317.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0600002148.rn.1 | TC0600000317.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0600002160.rn.1 | TC0600000317.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0600002157.rn.1 | TC0600000317.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0600002159.rn.1 | TC0600000317.rn.1 | RTA TC |
| Rat Expression Set 230 | 1370164_at | - | - |
| Rat Expression Set 230 | 1370164_PM_at | - | - |
| Rat Expression Set 230 | 1394857_PM_at | - | - |
| Rat Expression Set 230 | 1394763_PM_at | - | - |
| Rat Expression Set 230 | 1394763_at | - | - |
| Rat Expression Set 230 | 1394857_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10883305 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10883303 | - | - |
| Rat Genome U34 Set | D16478_g_at | - | - |
| Rat Genome U34 Set | X98225_s_at | - | - |
| Rat Genome U34 Set | D16478_at | - | - |
| Rat Genome U34 Set | D16478_at | - | - |
| Rat Genome U34 Set | D16478_g_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 22121368 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1376331 | - | - |

Hadha
Genomic Map