This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn00591868_m1
See other IGSF6 GE Assays ›
Gene Symbol:
Igsf6
Gene Name:
immunoglobulin superfamily, member 6
Gene Aliases:
DORA
Chromosome Location:
Chr.1: 190997029 - 191007503 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn00591868_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Igsf6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_133542.2 NP_598226.1 1 - 2 106 88
GenBank mRNA AJ223184.1 - 1 - 2 106 88

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0100006542.rn.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Igsf6

Panther Classification:

Molecular Function -

immunoglobulin

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P588738 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P588738 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P588738 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P588738 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P588738 - -
Matsumoto (Agilent Rat cDNA Microarray G4105A) 701096525 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 81178 - -
Agilent Rat cDNA Microarray (G4105A) 701096525 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10725400 - -
Affymetrix Rat Gene 1.0 ST Array 10725398 - -
Affymetrix Rat Gene 1.0 ST Array 10725401 - -
Affymetrix Rat Gene 1.0 ST Array 10725402 - -
Affymetrix Rat Gene 1.0 ST Array 10725403 - -
Affymetrix Rat Gene 1.0 ST Array 10725399 - -
Affymetrix Rat Gene 1.1 ST Array 10725397 - -
Rat Genome 230 2.0 Array 1387687_at - -
Rat Genome 230 2.0 Array 1387687_PM_at - -
Affymetrix HT Rat Focus Array 1387687_PM_at - -
Affymetrix HT Rat Focus Array 1387687_at - -
Affymetrix Rat Clariom S Assay PSR0100051388.rn.1 TC0100006542.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0100051389.rn.1 TC0100006542.rn.1 RTA TC
Rat Expression Set 230 1387687_at - -
Rat Expression Set 230 1387687_PM_at - -
Affymetrix Rat Gene 1.0 ST Array 10725397 - -
Rat Genome U34 Set AJ223184_at - -
Rat Genome U34 Set AJ223184_g_at - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 21140654 - -
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1360389 - -
RatRef-12 Expression BeadChip ILMN_59855 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-4sktr:80/100.66.76.202:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0