This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn00598400_m1
See other SLC1A5 GE Assays ›
Gene Symbol:
Slc1a5
Gene Name:
solute carrier family 1 (neutral amino acid transporter), member 5
Gene Aliases:
Asct2, H4-ASCT2, Slc1a7
Chromosome Location:
Chr.1: 78710686 - 78724789 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn00598400_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price 284.00
Your Price
Online offer: 266.65
266.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Slc1a5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_175758.3 NP_786934.1 3 - 4 1052 64
GenBank mRNA AJ132846.1 - 3 - 4 783 64
BC080242.1 - 3 - 4 1052 7100054 64

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0100000938.rn.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Slc1a5

Panther Classification:

Molecular Function -

primary active transporter

Gene Ontology Categories:

Function(s) Process(es)

amino acid transmembrane transport
dicarboxylic acid transport
extracellular amino acid transport
neutral amino acid transport
L-serine transport
amino acid transmembrane transporter activity
neutral amino acid transmembrane transporter activity
L-serine transmembrane transporter activity
glutamate binding
sodium:dicarboxylate symporter activity

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P111500 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 503865 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 228954 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 966565 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 947440 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 640238 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 504356 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 473612 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 462823 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 951435 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 325264 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 525800 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 40333 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P111500 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P111500 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P111500 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P829301 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P238222 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 37967 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 44508 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10704506 - -
Affymetrix Rat Gene 1.0 ST Array 10704508 - -
Affymetrix Rat Gene 1.0 ST Array 10704513 - -
Affymetrix Rat Gene 1.0 ST Array 10704507 - -
Affymetrix Rat Gene 1.0 ST Array 10704514 - -
Affymetrix Rat Gene 1.0 ST Array 10704516 - -
Affymetrix Rat Gene 1.0 ST Array 10704509 - -
Affymetrix Rat Gene 1.0 ST Array 10704511 - -
Affymetrix Rat Gene 1.0 ST Array 10704512 - -
Affymetrix Rat Gene 1.0 ST Array 10704515 - -
Affymetrix Rat Gene 1.1 ST Array 10704505 - -
Rat Genome 230 2.0 Array 1371040_at - -
Rat Genome 230 2.0 Array 1371040_PM_at - -
Affymetrix HT Rat Focus Array 1371040_PM_at - -
Affymetrix HT Rat Focus Array 1371040_at - -
Affymetrix Rat Clariom S Assay PSR0100006455.rn.1 TC0100000938.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0100006457.rn.1 TC0100000938.rn.1 RTA TC
Rat Expression Set 230 1371040_at - -
Rat Expression Set 230 1371040_PM_at - -
Affymetrix Rat Gene 1.0 ST Array 10704505 - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 21849990 - -
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1365597 - -
RatRef-12 Expression BeadChip ILMN_50868 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0