This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn00664607_m1
See other GLUD1 GE Assays ›
Gene Symbol:
Glud1
Gene Name:
glutamate dehydrogenase 1
Gene Aliases:
Ac2-281, Gdh1, Glud, Gludeha, MRG-2
Chromosome Location:
Chr.16: 10661911 - 10695557 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn00664607_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Glud1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_012570.2 NP_036702.1 11 - 12 1662 62
GenBank mRNA BC081841.1 - 11 - 12 1546 7107138 62
FQ210745.1 - 1 - 2 57 62
FQ214067.1 - 3 - 4 242 62
U95148.1 - 2 - 3 203 62
X14044.1 - 11 - 12 1552 62
X14223.1 - 11 - 12 1525 62

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC1600000147.rn.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Glud1

Panther Classification:

Molecular Function -

dehydrogenase

Gene Ontology Categories:

Function(s) Process(es)

glutamate biosynthetic process
glutamate catabolic process
glutamine metabolic process
long-term memory
response to aluminum ion
substantia nigra development
positive regulation of insulin secretion
oxidation-reduction process
tricarboxylic acid metabolic process
glutamate dehydrogenase (NAD+) activity
glutamate dehydrogenase [NAD(P)+] activity
ATP binding
GTP binding
enzyme binding
ADP binding
NAD+ binding
leucine binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR1600001677.rn.1 TC1600000147.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-cjlz8:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0