This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn00664980_m1
See other SRD5A1 GE Assays ›
Gene Symbol:
Srd5a1
Gene Name:
steroid-5-alpha-reductase, alpha polypeptide 1 (3-oxo-5 alpha-steroid delta 4-dehydrogenase alpha 1)
Gene Aliases:
S5AR 1
Chromosome Location:
Chr.1: 36320504 - 36354694 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn00664980_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Srd5a1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_017070.3 NP_058766.2 856 118
GenBank mRNA BC127454.1 - 857 7313862 118
EF091857.1 - 342 118
J05035.1 - 709 118
S81448.1 - 729 118

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0100000376.rn.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Srd5a1

Panther Classification:

Molecular Function -

dehydrogenase

Gene Ontology Categories:

Function(s) Process(es)

urogenital system development
liver development
steroid biosynthetic process
androgen biosynthetic process
androgen metabolic process
male gonad development
cellular response to starvation
response to organic cyclic compound
response to muscle activity
diterpenoid metabolic process
spinal cord development
hippocampus development
thalamus development
hypothalamus development
pituitary gland development
cerebral cortex development
cell differentiation
male genitalia development
female genitalia development
response to follicle-stimulating hormone
response to estradiol
cellular response to insulin stimulus
response to testosterone
serotonin metabolic process
progesterone metabolic process
response to drug
circadian sleep/wake cycle, REM sleep
response to estrogen
male sex differentiation
oxidation-reduction process
bone development
response to growth hormone
response to fungicide
cellular response to cAMP
cellular response to growth factor stimulus
cellular response to estradiol stimulus
cellular response to testosterone stimulus
cellular response to organic cyclic compound
cellular response to dexamethasone stimulus
cellular response to epinephrine stimulus
3-oxo-5-alpha-steroid 4-dehydrogenase activity
amide binding
cholestenone 5-alpha-reductase activity
NADPH binding

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P519871 - -
Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P182431 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 410340 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 879431 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 789385 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 419465 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 969050 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 31253 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 865771 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 945447 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 117393 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 645189 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 616800 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 472362 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 720645 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 611400 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 115967 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 62995 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 140168 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 546463 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 333286 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 886262 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 727606 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 883203 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 508063 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 717721 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 968863 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 918315 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 91670 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 56803 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 35594 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 139025 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P182431 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P519871 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P182431 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P519871 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P519871 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P182431 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P519871 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P182431 - -
Matsumoto (Agilent Rat cDNA Microarray G4105A) 701858004 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 49986 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 46444 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 80236 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 90571 - -
Agilent Rat cDNA Microarray (G4105A) 701858004 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10793315 - -
Affymetrix Rat Gene 1.0 ST Array 10793312 - -
Affymetrix Rat Gene 1.0 ST Array 10793311 - -
Affymetrix Rat Gene 1.0 ST Array 10793313 - -
Affymetrix Rat Gene 1.0 ST Array 10793316 - -
Affymetrix Rat Gene 1.1 ST Array 10793310 - -
Rat Genome 230 2.0 Array 1370420_PM_at - -
Rat Genome 230 2.0 Array 1370420_at - -
Affymetrix HT Rat Focus Array 1370420_PM_at - -
Affymetrix HT Rat Focus Array 1370420_at - -
Affymetrix Rat Clariom S Assay PSR0100002250.rn.1 TC0100000376.rn.1 RTA TC
Rat Expression Set 230 1370420_at - -
Rat Expression Set 230 1370420_PM_at - -
Affymetrix Rat Gene 1.0 ST Array 10793310 - -
Rat Genome U34 Set J05035_g_at - -
Rat Genome U34 Set S81448_s_at - -
Rat Genome U34 Set J05035_at - -
Rat Genome U34 Set J05035_at - -
Rat Genome U34 Set J05035_g_at - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 20857839 - -
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1364151 - -
RatRef-12 Expression BeadChip ILMN_62683 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-5h84w:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0