This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn00666277_g1
See other FABP9 GE Assays ›
Gene Symbol:
Fabp9
Gene Name:
fatty acid binding protein 9, testis
Gene Aliases:
Tlbp
Chromosome Location:
Chr.2: 93803503 - 93807069 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn00666277_g1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Fabp9
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_022854.1 NP_074045.1 3 - 4 386 91
GenBank mRNA BC078817.1 - 3 - 4 387 7133719 91
U07870.1 - 3 - 4 372 91
U09022.1 - 3 - 4 386 91

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0200000779.rn.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Fabp9

Gene Ontology Categories:

Function(s) Process(es)

acrosome assembly
transport
transporter activity
lipid binding

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P143547 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 470251 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 917096 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 74278 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P143547 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P143547 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P143547 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12321 - -
Matsumoto (Agilent Rat cDNA Microarray G4105A) 700489724 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 52861 - -
Agilent Rat cDNA Microarray (G4105A) 700489724 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10814296 - -
Affymetrix Rat Gene 1.0 ST Array 10814295 - -
Affymetrix Rat Gene 1.0 ST Array 10814298 - -
Affymetrix Rat Gene 1.0 ST Array 10814297 - -
Affymetrix Rat Gene 1.1 ST Array 10814294 - -
Rat Genome 230 2.0 Array 1368630_at - -
Rat Genome 230 2.0 Array 1368630_PM_at - -
Affymetrix HT Rat Focus Array 1368630_at - -
Affymetrix HT Rat Focus Array 1368630_PM_at - -
Affymetrix Rat Clariom S Assay PSR0200004821.rn.1 TC0200000779.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0200004822.rn.1 TC0200000779.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0200004823.rn.1 TC0200000779.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0200004824.rn.1 TC0200000779.rn.1 RTA TC
Rat Expression Set 230 1368630_PM_at - -
Rat Expression Set 230 1368630_at - -
Affymetrix Rat Gene 1.0 ST Array 10814294 - -
Rat Genome U34 Set U09022_at - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 21594793 - -
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1349079 - -
RatRef-12 Expression BeadChip ILMN_51777 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0