This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn00668380_g1
See other PLA2G2A GE Assays ›
Gene Symbol:
Pla2g2a
Gene Name:
phospholipase A2, group IIA (platelets, synovial fluid)
Gene Aliases:
sPLA2
Chromosome Location:
Chr.5: 157282650 - 157285295 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn00668380_g1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pla2g2a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_031598.3 NP_113786.3 3 - 4 245 85
XM_006239146.2 XP_006239208.1 4 - 5 411 85
GenBank mRNA AF365363.1 - 2 - 3 189 85
AY651027.1 - 2 - 3 189 85
D00523.1 - 3 - 4 229 85
FQ227448.1 - 3 - 4 245 85
FQ234749.1 - 4 - 5 343 85
M25148.1 - 3 - 4 245 85
X74364.1 - 1 - 2 125 85

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0500001759.rn.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Pla2g2a

Panther Classification:

Molecular Function -

phospholipase

Gene Ontology Categories:

Function(s) Process(es)

phospholipid metabolic process
negative regulation of cell proliferation
lipid catabolic process
somatic stem cell population maintenance
regulation of cell proliferation
phosphatidic acid metabolic process
regulation of epithelial cell proliferation
negative regulation of epithelial cell proliferation
phospholipase A2 activity
calcium ion binding
phospholipid binding
calcium-dependent phospholipase A2 activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0500013087.rn.1 TC0500001759.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0