This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn00669387_m1
See other APBA1 GE Assays ›
Gene Symbol:
Apba1
Gene Name:
amyloid beta (A4) precursor protein-binding, family A, member 1
Gene Aliases:
Mint1
Chromosome Location:
Chr.1: 241594565 - 241799456 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn00669387_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Apba1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_031779.2 NP_113967.1 12 - 13 2791 59
XM_006231185.2 XP_006231247.1 10 - 11 2891 59
XM_008760312.1 XP_008758534.1 11 - 12 2924 59
GenBank mRNA AF029105.1 - 3011 59

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0100003388.rn.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Apba1

Panther Classification:

Molecular Function -

membrane trafficking regulatory protein

Gene Ontology Categories:

Function(s) Process(es)

in utero embryonic development
intracellular protein transport
chemical synaptic transmission
locomotory behavior
regulation of gene expression
glutamate secretion
gamma-aminobutyric acid secretion
synaptic vesicle exocytosis
multicellular organism growth
beta-amyloid binding
protein binding
phosphatidylinositol-4,5-bisphosphate binding
PDZ domain binding
protein complex binding

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P075234 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 863420 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 512268 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 261733 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 339402 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 880676 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 6247 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 588543 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 85362 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 597335 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 153322 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 879639 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 322083 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 38935 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 252349 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 239137 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 726233 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 792381 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 968836 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 65752 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 662619 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 153565 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 330259 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 730412 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 690888 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 522912 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 160464 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 279379 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 356592 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 940469 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 304295 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 194220 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 655782 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 308095 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 838325 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 830382 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 899617 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 51851 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 611103 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 832435 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 533050 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 94263 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 455926 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 228201 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 582111 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 364710 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 708177 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 860323 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 690618 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 599966 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 2670 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 659281 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 651965 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 289064 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 831325 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 769546 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 453406 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 663433 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 838589 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 445305 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 420638 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 238574 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 210590 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 939817 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 36218 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 847610 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 944849 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 894684 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 812068 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 192707 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 113648 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 458041 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 451830 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 333529 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 151487 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 501347 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 411519 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 515927 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 390748 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 623090 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 93124 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 727574 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 567859 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 254200 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 291874 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 802500 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 498755 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 938294 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 421498 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 487203 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 905058 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 157611 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 429924 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 237504 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 245317 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 776000 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 188508 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 306457 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 898495 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 21480 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 36961 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 579899 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 382765 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 174571 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 507584 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 676858 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 755378 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 147346 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 607675 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 130154 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 462322 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 366130 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 131175 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 518447 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 88742 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 630563 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 407377 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 105111 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 399422 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 254601 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 576608 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 706057 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 818182 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 133782 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 373608 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 489043 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 565169 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 19024 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 673578 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 2460 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 526365 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 701595 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 690216 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 664051 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 294992 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 488699 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 461204 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 730764 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 319913 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 726165 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 141085 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 130507 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 866609 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 171343 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 808939 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 840012 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 491702 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 630575 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 70695 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 145670 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 83689 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 912411 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 33633 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 118485 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 138188 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 149085 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 119431 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 85735 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 104151 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 116315 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 169930 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 24704 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 121277 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 41625 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 172660 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 80624 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 65955 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 145058 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 28820 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 130861 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 156169 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 115586 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 123011 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 127961 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 177087 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P075234 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P075234 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P075234 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P649121 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 47691 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 29297 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 76661 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 70874 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 48390 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 10357 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 16680 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 42972 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 104348 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 82170 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 54743 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 103728 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10714443 - -
Affymetrix Rat Gene 1.0 ST Array 10714442 - -
Affymetrix Rat Gene 1.0 ST Array 10714436 - -
Affymetrix Rat Gene 1.0 ST Array 10714438 - -
Affymetrix Rat Gene 1.0 ST Array 10714444 - -
Affymetrix Rat Gene 1.0 ST Array 10714434 - -
Affymetrix Rat Gene 1.0 ST Array 10714445 - -
Affymetrix Rat Gene 1.0 ST Array 10714440 - -
Affymetrix Rat Gene 1.0 ST Array 10714441 - -
Affymetrix Rat Gene 1.0 ST Array 10714433 - -
Affymetrix Rat Gene 1.0 ST Array 10714435 - -
Affymetrix Rat Gene 1.0 ST Array 10714437 - -
Affymetrix Rat Gene 1.0 ST Array 10714439 - -
Affymetrix Rat Gene 1.1 ST Array 10714432 - -
Rat Genome 230 2.0 Array 1387425_PM_at - -
Rat Genome 230 2.0 Array 1387425_at - -
Affymetrix HT Rat Focus Array 1387425_at - -
Affymetrix HT Rat Focus Array 1387425_PM_at - -
Affymetrix Rat Clariom S Assay PSR0100027514.rn.1 TC0100003388.rn.1 RTA TC
Rat Expression Set 230 1387425_at - -
Rat Expression Set 230 1387425_PM_at - -
Affymetrix Rat Gene 1.0 ST Array 10714432 - -
Rat Genome U34 Set AF029105_at - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 21991008 - -
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1351230 - -
RatRef-12 Expression BeadChip ILMN_49871 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0