This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn00672490_g1
See other POLR2G GE Assays ›
Gene Symbol:
Polr2g
Gene Name:
polymerase (RNA) II (DNA directed) polypeptide G
Gene Aliases:
Rpb7
Chromosome Location:
Chr.1: 224991250 - 224998355 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn00672490_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Polr2g
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_053948.4 NP_446400.1 7 - 8 572 140
XM_006230973.1 XP_006231035.1 9 - 10 857 140
GenBank mRNA BC060550.1 - 7 - 8 528 6890594 140
BC060595.1 - 9 - 10 866 6888231 140
FQ223906.1 - 9 - 10 862 140
FQ230390.1 - 7 - 8 535 140
FQ232354.1 - 7 - 8 576 140
Z71925.1 - 7 - 8 573 140

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0100007220.rn.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Polr2g

Panther Classification:

Molecular Function -

DNA-directed RNA polymerase

Gene Ontology Categories:

Function(s) Process(es)

nuclear-transcribed mRNA catabolic process, exonucleolytic
transcription from RNA polymerase II promoter
transcription initiation from RNA polymerase II promoter
apoptotic process
positive regulation of translational initiation
positive regulation of nuclear-transcribed mRNA poly(A) tail shortening
molecular_function
single-stranded DNA binding
single-stranded RNA binding
DNA-directed RNA polymerase activity
translation initiation factor binding

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P035895 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 683213 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 210264 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 57851 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 474198 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 738294 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 175540 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P035895 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P035895 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P035895 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P524690 - -
Matsumoto (Agilent Rat cDNA Microarray G4105A) 700544084 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 23457 - -
Agilent Rat cDNA Microarray (G4105A) 700544084 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10728539 - -
Affymetrix Rat Gene 1.0 ST Array 10728542 - -
Affymetrix Rat Gene 1.0 ST Array 10728544 - -
Affymetrix Rat Gene 1.0 ST Array 10728548 - -
Affymetrix Rat Gene 1.0 ST Array 10728547 - -
Affymetrix Rat Gene 1.0 ST Array 10728545 - -
Affymetrix Rat Gene 1.0 ST Array 10728540 - -
Affymetrix Rat Gene 1.0 ST Array 10728546 - -
Affymetrix Rat Gene 1.0 ST Array 10728541 - -
Affymetrix Rat Gene 1.0 ST Array 10728543 - -
Affymetrix Rat Gene 1.1 ST Array 10728538 - -
Rat Genome 230 2.0 Array 1367769_PM_at - -
Rat Genome 230 2.0 Array 1367769_at - -
Affymetrix HT Rat Focus Array 1367769_PM_at - -
Affymetrix HT Rat Focus Array 1367769_at - -
Affymetrix Rat Clariom S Assay PSR0100058263.rn.1 TC0100007220.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0100058262.rn.1 TC0100007220.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0100058264.rn.1 TC0100007220.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0100058266.rn.1 TC0100007220.rn.1 RTA TC
Rat Expression Set 230 1367769_PM_at - -
Rat Expression Set 230 1367769_at - -
Affymetrix Rat Gene 1.0 ST Array 10728538 - -
Rat Genome U34 Set Z71925_at - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 21012534 - -
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1356866 - -
RatRef-12 Expression BeadChip ILMN_69902 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0