This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn00672503_m1
See other EIF2B4 GE Assays ›
Gene Symbol:
Eif2b4
Gene Name:
eukaryotic translation initiation factor 2B, subunit 4 delta
Gene Aliases:
Eif2b
Chromosome Location:
Chr.6: 26546917 - 26552474 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn00672503_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Eif2b4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_053950.1 NP_446402.1 11 - 12 1197 61
GenBank mRNA FQ233756.1 - 6 - 7 669 61
Z48225.1 - 11 - 12 1197 61

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0600000286.rn.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Eif2b4

Panther Classification:

Molecular Function -

translation initiation factor

Gene Ontology Categories:

Function(s) Process(es)

ovarian follicle development
translational initiation
regulation of translation
response to heat
response to glucose
oligodendrocyte development
L-methionine biosynthetic process from methylthioadenosine
negative regulation of translational initiation in response to stress
myelination
response to peptide hormone
positive regulation of GTPase activity
cellular metabolic process
translation initiation factor activity
guanyl-nucleotide exchange factor activity
translation initiation factor binding
S-methyl-5-thioribose-1-phosphate isomerase activity

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0600001822.rn.1 TC0600000286.rn.1 RTA TC
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 21725490 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0