This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn00672963_m1
See other IL1RN GE Assays ›
Gene Symbol:
Il1rn
Gene Name:
interleukin 1 receptor antagonist
Gene Aliases:
IL-1ra
Chromosome Location:
Chr.3: 1450536 - 1468624 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn00672963_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Il1rn
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_022194.2 NP_071530.1 3 - 4 330 78
XM_006233636.2 XP_006233698.1 4 - 5 2573 78
XM_006233637.2 XP_006233699.1 4 - 5 360 78
XM_006233638.2 XP_006233700.1 3 - 4 260 78
GenBank mRNA BC070930.1 - 3 - 4 324 78
CK364274.1 - 3 - 4 354 78

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0300000017.rn.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Il1rn

Panther Classification:

Molecular Function -

interleukin superfamily

Gene Ontology Categories:

Function(s) Process(es)

fever generation
negative regulation of cytokine-mediated signaling pathway
chronic inflammatory response to antigenic stimulus
lipid metabolic process
acute-phase response
female pregnancy
memory
negative regulation of signal transduction
response to organonitrogen compound
negative regulation of glutamate secretion
response to organic cyclic compound
response to activity
cytokine-mediated signaling pathway
sensory perception of pain
carboxylic acid metabolic process
insulin secretion
negative regulation of cell migration
response to lipopolysaccharide
negative regulation of heterotypic cell-cell adhesion
response to drug
response to starvation
negative regulation of apoptotic process
positive regulation of JUN kinase activity
negative regulation of membrane potential
response to glucocorticoid
response to interleukin-1
response to interleukin-4
cellular response to norepinephrine stimulus
negative regulation of interleukin-1-mediated signaling pathway
cytokine activity
interleukin-1 receptor binding
interleukin-1, Type I receptor binding
interleukin-1, Type II receptor binding
interleukin-1 receptor antagonist activity
interleukin-1 Type I receptor antagonist activity
interleukin-1 Type II receptor antagonist activity

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P462661 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 70146 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 201960 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 98995 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P462661 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P462661 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P462661 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P112890 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P462661 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 90607 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10834111 - -
Affymetrix Rat Gene 1.0 ST Array 10834114 - -
Affymetrix Rat Gene 1.0 ST Array 10834113 - -
Affymetrix Rat Gene 1.0 ST Array 10834112 - -
Affymetrix Rat Gene 1.1 ST Array 10834109 - -
Rat Genome 230 2.0 Array 1387835_PM_at - -
Rat Genome 230 2.0 Array 1387835_at - -
Affymetrix HT Rat Focus Array 1387835_PM_at - -
Affymetrix HT Rat Focus Array 1387835_at - -
Affymetrix Rat Clariom S Assay PSR0300000065.rn.1 TC0300000017.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0300000064.rn.1 TC0300000017.rn.1 RTA TC
Rat Expression Set 230 1387835_PM_at - -
Rat Expression Set 230 1387835_at - -
Affymetrix Rat Gene 1.0 ST Array 10834109 - -
Rat Genome U34 Set M63101cds_at - -
Rat Genome U34 Set M63101cds_at - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 21130456 - -
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1361277 - -
RatRef-12 Expression BeadChip ILMN_50105 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l2rv2:80/100.66.78.26:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0