This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn00673915_m1
See other GPX3 GE Assays ›
Gene Symbol:
Gpx3
Gene Name:
glutathione peroxidase 3
Gene Aliases:
GPx-3, GPx-P, GSHPx-3, GSHPx-P, Gpxp
Chromosome Location:
Chr.10: 40247436 - 40255423 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn00673915_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gpx3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_022525.3 NP_071970.2 2 - 3 309 70
GenBank mRNA BC062227.1 - 2 - 3 285 6921653 70
CK480346.1 - 2 - 3 309 70
CV079715.1 - 1 - 2 48 70
D00680.1 - 2 - 3 374 70
FQ221037.1 - 2 - 3 281 70
FQ221279.1 - 2 - 3 262 70
FQ221454.1 - 2 - 3 281 70
FQ228545.1 - 2 - 3 335 70
FQ228679.1 - 2 - 3 281 70
FQ228689.1 - 2 - 3 281 70
FQ228690.1 - 2 - 3 294 70
FQ228694.1 - 2 - 3 261 70
FQ228705.1 - 2 - 3 279 70
FQ228712.1 - 2 - 3 281 70
FQ228719.1 - 2 - 3 281 70
FQ228737.1 - 2 - 3 281 70
FQ228751.1 - 2 - 3 281 70
FQ228788.1 - 2 - 3 281 70
FQ228802.1 - 2 - 3 281 70
FQ228804.1 - 2 - 3 281 70
FQ228807.1 - 2 - 3 282 70
FQ228808.1 - 2 - 3 281 70
FQ228824.1 - 2 - 3 281 70
FQ228834.1 - 2 - 3 283 70
FQ228835.1 - 2 - 3 281 70
FQ228845.1 - 2 - 3 281 70
FQ228866.1 - 2 - 3 281 70
FQ228878.1 - 2 - 3 281 70
FQ228886.1 - 2 - 3 281 70
FQ228899.1 - 2 - 3 281 70
FQ228911.1 - 2 - 3 281 70
FQ228914.1 - 2 - 3 281 70
FQ228921.1 - 2 - 3 281 70
FQ228934.1 - 2 - 3 283 70
FQ228945.1 - 2 - 3 281 70
FQ228947.1 - 2 - 3 281 70
FQ228960.1 - 2 - 3 281 70
FQ228965.1 - 2 - 3 281 70
FQ228968.1 - 2 - 3 281 70
FQ228969.1 - 2 - 3 280 70
FQ228970.1 - 2 - 3 281 70
FQ228976.1 - 2 - 3 281 70
FQ229118.1 - 2 - 3 281 70
FQ229125.1 - 2 - 3 281 70

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC1000000614.rn.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gpx3

Panther Classification:

Molecular Function -

peroxidase

Gene Ontology Categories:

Function(s) Process(es)

response to molecule of fungal origin
glutathione metabolic process
response to oxidative stress
female pregnancy
response to organic substance
response to selenium ion
response to organic cyclic compound
response to drug
hydrogen peroxide catabolic process
protein homotetramerization
response to corticosterone
oxidation-reduction process
glutathione peroxidase activity
selenium binding
glutathione binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0