This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn00680648_m1
See other CEACAM3 GE Assays ›
Gene Symbol:
Ceacam3
Gene Name:
carcinoembryonic antigen-related cell adhesion molecule 3
Gene Aliases:
CGM4AA, Cea, Ceacam5, Cear, Cgm1, Cgm4, RATCEAA, RATCGM4AA
Chromosome Location:
Chr.1: 79122593 - 79135127 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn00680648_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ceacam3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_012702.2 NP_036834.2 2 - 3 555 101
GenBank mRNA BC098921.1 - 2 - 3 555 7441743 101
M32474.1 - 2 - 3 540 101

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0100000958.rn.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ceacam3

Panther Classification:

Molecular Function -

immunoglobulin receptor superfamily

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0100006629.rn.1 TC0100000958.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0100006630.rn.1 TC0100000958.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0100006631.rn.1 TC0100000958.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-wtj7v:80/100.66.79.197:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline