This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn00681293_g1
See other FOSL1 GE Assays ›
Gene Symbol:
Fosl1
Gene Name:
fos-like antigen 1
Gene Aliases:
Fra-1, fra-1
Chromosome Location:
Chr.1: 220826560 - 220835066 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn00681293_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Fosl1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_012953.1 NP_037085.1 3 - 4 638 71
GenBank mRNA BC097342.1 - 3 - 4 617 7459558 71
M19651.1 - 3 - 4 638 71

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0100003054.rn.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Fosl1

Panther Classification:

Molecular Function -

basic leucine zipper transcription factor

Gene Ontology Categories:

Function(s) Process(es)

in utero embryonic development
vitellogenesis
female pregnancy
learning
negative regulation of cell proliferation
response to mechanical stimulus
response to gravity
response to organic cyclic compound
cellular response to extracellular stimulus
response to progesterone
response to cytokine
response to drug
response to hydrogen peroxide
positive regulation of apoptotic process
positive regulation of cell cycle
positive regulation of transcription from RNA polymerase II promoter
positive regulation of sequence-specific DNA binding transcription factor activity
response to corticosterone
response to cAMP
placenta blood vessel development
RNA polymerase II regulatory region sequence-specific DNA binding
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
DNA binding

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P123118 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 596131 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 253886 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 298446 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 412206 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 233886 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 903012 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 823461 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 125763 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 100321 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P123118 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P123118 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P123118 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P693821 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 100251 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 88705 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10713047 - -
Affymetrix Rat Gene 1.0 ST Array 10713049 - -
Affymetrix Rat Gene 1.0 ST Array 10713046 - -
Affymetrix Rat Gene 1.0 ST Array 10713048 - -
Affymetrix Rat Gene 1.1 ST Array 10713045 - -
Rat Genome 230 2.0 Array 1368489_PM_at - -
Rat Genome 230 2.0 Array 1368489_at - -
Affymetrix HT Rat Focus Array 1368489_PM_at - -
Affymetrix HT Rat Focus Array 1368489_at - -
Affymetrix Rat Clariom S Assay PSR0100024737.rn.1 TC0100003054.rn.1 RTA TC
Rat Expression Set 230 1368489_at - -
Rat Expression Set 230 1368489_PM_at - -
Affymetrix Rat Gene 1.0 ST Array 10713045 - -
Rat Genome U34 Set M19651_at - -
Rat Genome U34 Set M19651_at - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 21305806 - -
Array Name Probe IDs Targets Target Type
RatRef-12 Expression BeadChip ILMN_54965 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline