This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn00683893_m1
See other ATP5F1 GE Assays ›
Gene Symbol:
Atp5f1 LOC100911417
Gene Name:
ATP synthase, H+ transporting, mitochondrial Fo complex, subunit B1
ATP synthase subunit b, mitochondrial-like
Gene Aliases:
-
-
Chromosome Location:
Chr.2: 208566385 - 208577147 on Build Rnor_6.0
Chromosome Location:
Chr.2: 208408890 - 208420159 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
-
Assay ID Rn00683893_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Atp5f1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_134365.2 NP_599192.1 2 - 3 220 73
GenBank mRNA BC063808.1 - 2 - 3 161 6920021 73
FQ209675.1 - 2 - 3 119 73
FQ212544.1 - 2 - 3 156 73
FQ214336.1 - 2 - 3 148 73
FQ214353.1 - 2 - 3 182 73
FQ214371.1 - 2 - 3 156 73
FQ214449.1 - 2 - 3 119 73
FQ214672.1 - 2 - 3 155 73
FQ214784.1 - 2 - 3 154 73
FQ215244.1 - 2 - 3 119 73
FQ215436.1 - 2 - 3 119 73
FQ215480.1 - 2 - 3 119 73
FQ215788.1 - 2 - 3 155 73
FQ215823.1 - 2 - 3 172 73
FQ216062.1 - 2 - 3 121 73
FQ216247.1 - 2 - 3 105 73
FQ216597.1 - 2 - 3 111 73
FQ216773.1 - 2 - 3 119 73
FQ216829.1 - 2 - 3 119 73
FQ216922.1 - 2 - 3 119 73
FQ218701.1 - 2 - 3 155 73
FQ219899.1 - 2 - 3 148 73
FQ223652.1 - 2 - 3 119 73
FQ223683.1 - 2 - 3 145 73
FQ229261.1 - 2 - 3 179 73
FQ229976.1 - 2 - 3 156 73
FQ232356.1 - 2 - 3 156 73
M35052.1 - 2 - 3 139 73
Gene Symbol: LOC100911417
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_003749372.2 XP_003749420.1 215 73

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Rattus norvegicus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Atp5f1
Click here to find siRNAs for Gene LOC100911417

Panther Classification:

Molecular Function -

ATP synthase

Gene Ontology Categories:

Function(s) Process(es)

ATP synthesis coupled proton transport
ATP metabolic process
substantia nigra development
proton-transporting ATP synthase activity, rotational mechanism
ATPase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0200030430.rn.1 TC0200004408.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-gznm4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0